1-(2-Bromethoxy)-3-nitrobenzol 97%

Lieferant: Thermo Scientific
Achtung

385260050 385260250
ACRO385260050EA 39.5 EUR
ACRO385260050 ACRO385260250
1-(2-Bromethoxy)-3-nitrobenzol 97%
1-(2-Bromethoxy)-3-nitrobenzol
Formel: C₈H₈BrNO₃
Molecular Weight: 246,06 g/mol
Siedepunkt: 140…145 °C (2 mmHg)
Schmelzpunkt: 39 °C
MDL: MFCD07783648
CAS-Nummer: 13831-59-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow-green to brown Low melting solid
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR