1-Brom-3,5-dimethoxybenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

569313-25G 569313-5G
SIAL569313-25GEA 113 EUR
SIAL569313-25G SIAL569313-5G
1-Brom-3,5-dimethoxybenzol, Sigma-Aldrich®
1-Brom-3,5-dimethoxybenzol
Formel: C₈H₉BrO₂
Molecular Weight: 217,06 g/mol
MDL: MFCD06200685
CAS-Nummer: 20469-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR