3-Bromthioanisol
Lieferant: FLUOROCHEM
Synonyme:
1-Brom-3-(methylthio)benzol, 3-Bromphenylmethylsulfid
003661-25G
003661-100G
FLUO003661-25GEA
165
EUR
FLUO003661-25G
FLUO003661-100G
3-Bromthioanisol
3-Bromthioanisol
Formel:
C₇H₇BrS Molecular Weight: 203,1 g/mol |
MDL:
MFCD00041395 CAS-Nummer: 33733-73-2 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...