3-Brombenzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3-Brombenzolsulfonsäurechlorid, 3-Brombenzolsulfochlorid
003578-5G
003578-25G
003578-100G
FLUO003578-5GEA
36.8
EUR
FLUO003578-5G
FLUO003578-25G
FLUO003578-100G
3-Brombenzolsulfonylchlorid
3-Brombenzolsulfonylchlorid
Formel:
C₆H₄BrClO₂S Molecular Weight: 255,52 g/mol Siedepunkt: 95…95,5 °C (0.8 mmHg) Schmelzpunkt: 31…33 °C Dichte: 1,773 g/cm³ (20 °C) |
MDL:
MFCD00052313 CAS-Nummer: 2905-24-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...