2-Cyanbenzaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Formylbenzonitrile

H29027.06 H29027.03
H29027.06EA 136 EUR
H29027.06 H29027.03
2-Cyanbenzaldehyd ≥98%
2-Cyanbenzaldehyd

Purity: 98% CAS No.: 7468-67-9 Molecular Formula: C8H5NO Formula Weight: 131.13 Odour: Characteristic Melting Point: 102-106° MDL No.: MFCD00017503

Formel: C₈H₅NO
Molecular Weight: 131,13 g/mol
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017503
CAS-Nummer: 7468-67-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR