2-Cyanbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Formylbenzonitrile

M-Clarity™ Qualitätsstufe = MQ100

349089-1G 349089-5G
SIAL349089-1GEA 30.2 EUR
SIAL349089-1G SIAL349089-5G
2-Cyanbenzaldehyd, Sigma-Aldrich®
2-Cyanbenzaldehyd
Formel: C₈H₅NO
Molecular Weight: 131,13 g/mol
MDL: MFCD00017503
CAS-Nummer: 7468-67-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR