3-Cyanbenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Formylbenzonitril

A15943.03 A15943.06 A15943.14
A15943.03EA 18.8 EUR
A15943.03 A15943.06 A15943.14
3-Cyanbenzaldehyd ≥97%
3-Cyanbenzaldehyd

Purity: 97% CAS No.: 24964-64-5 Molecular Formula: C8H5NO Formula Weight: 131.13 Odour: Characteristic Melting Point: 75-79° Boiling Point: 209-210° Beilstein Reference: 2205751 MDL No.: MFCD00003344

Formel: C₈H₅NO
Molecular Weight: 131,13 g/mol
Siedepunkt: 209…210 °C
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003344
CAS-Nummer: 24964-64-5
EINECS: 246-549-1
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR