6-Nitroveratrumaldehyd 96%
Lieferant: Thermo Scientific
Synonyme:
4,5-Dimethoxy-2-nitrobenzaldehyd, 3,4-Dimethoxy-6-nitrobenzaldehyd
A14415.06
A14415.09
A14415.14
A14415.06EA
55.8
EUR
A14415.06
A14415.09
A14415.14
6-Nitroveratrumaldehyd 96%
6-Nitroveratrumaldehyd
Purity: 96%
CAS No.: 20357-25-9
Molecular Formula: C9H9NO5
Formula Weight: 211.17
Melting Point: 131-135°
Beilstein Reference: 395599
Formel:
C₉H₉NO₅ Molecular Weight: 211,17 g/mol Schmelzpunkt: 131…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007134 CAS-Nummer: 20357-25-9 EINECS: 243-762-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...