3-Nitrosalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Hydroxy-3-nitrobenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

527424-1G 527424-5G
SIAL527424-1GEA 33.4 EUR
SIAL527424-1G SIAL527424-5G
3-Nitrosalicylaldehyd, Sigma-Aldrich®
3-Nitrosalicylaldehyd
Formel: C₇H₅NO₄
Molecular Weight: 167,12 g/mol
MDL: MFCD00041874
CAS-Nummer: 5274-70-4
EINECS: 226-098-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR