2,3,5-Trichlorphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,3,5-Trichlorobenzeneboronic acid, Dihydroxy(2,3,5-trichlorophenyl)borane, (2,3,5-Trichlorophenyl)metaboric acid
L17511.06
L17511.03
L17511.06EA
200
EUR
L17511.06
L17511.03
2,3,5-Trichlorphenylboronsäure ≥98%
2,3,5-Trichlorphenylboronsäure
Purity: 98%
CAS No.: 212779-19-6
Molecular Formula: C6H4BCl3O2
Formula Weight: 225.27
Melting Point: ca 245°
MDL No.: MFCD01630856
Formel:
C₆H₄BCl₃O₂ Molecular Weight: 225,26 g/mol Schmelzpunkt: ∼245 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01630856 CAS-Nummer: 212779-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...