2,6-Dichlorphenylboronsäure

Lieferant: CHEMPUR
Achtung

Synonyme: (2,6-Dichlorphenyl)metaborsäure, 2,6-Dichlorbenzolboronsäure, Dihydroxy(2,6-dichlorphenyl)boran

FL11028.POR 011028-5G 011028-25G
CHMPFL11028.POREA 0 EUR
CHMPFL11028.POR FLUO011028-5G FLUO011028-25G
2,6-Dichlorphenylboronsäure
2,6-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190.82 g/mol
MDL: MFCD00064869
CAS-Nummer: 73852-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR