2-Chlorphenylboronsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
2-Chlorbenzolboronsäure, Dihydroxy(2-chlorphenyl)boran, (2-Chlorphenyl)metaborsäure
FL10984.POR
010984-25G
010984-5G
CHMPFL10984.POREA
0
EUR
CHMPFL10984.POR
FLUO010984-25G
FLUO010984-5G
2-Chlorphenylboronsäure
2-Chlorphenylboronsäure
Formel:
C₆H₆BClO₂ Molecular Weight: 156.38 g/mol |
MDL:
MFCD00674012 CAS-Nummer: 3900-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...