3,5-Dichlorphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

445207-25G 445207-5G
SIAL445207-25GEA 138 EUR
SIAL445207-25G SIAL445207-5G
3,5-Dichlorphenylboronsäure, Sigma-Aldrich®
3,5-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190,82 g/mol
MDL: MFCD00051935
CAS-Nummer: 67492-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR