3-Chlorphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(3-Chlorphenyl)metaborsäure, Dihydroxy(3-chlorphenyl)boran, 3-Chlorbenzolboronsäure
AC35900DA
AC35900.50
359000050
359000010
MAYBAC35900DAEA
0
EUR
MAYBAC35900DA
MAYBAC35900.50
ACRO359000050
ACRO359000010
3-Chlorphenylboronsäure 97%
3-Chlorphenylboronsäure
Formel:
C₆H₆BClO₂ Molecular Weight: 156.38 g/mol Schmelzpunkt: 185…189 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00161354 CAS-Nummer: 63503-60-6 |
Spezifikation Testergebnisse
Appearance | White to yellow-green Crystalline powder or needles |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...