3-Chlorphenylboronsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
(3-Chlorphenyl)metaborsäure, Dihydroxy(3-chlorphenyl)boran, 3-Chlorbenzolboronsäure
FL10985.POR
010985-25G
010985-5G
CHMPFL10985.POREA
0
EUR
CHMPFL10985.POR
FLUO010985-25G
FLUO010985-5G
3-Chlorphenylboronsäure
3-Chlorphenylboronsäure
Formel:
C₆H₆BClO₂ Molecular Weight: 156,38 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161354 CAS-Nummer: 63503-60-6 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...