4-Chlorphenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chlorbenzolboronsäure, Dihydroxy(4-chlorphenyl)boran, (4-Chlorphenyl)metaborsäure

A15657.03 A15657.06 A15657.14 AB109878.0005
A15657.03EA 27.8 EUR
A15657.03 A15657.06 A15657.14 ABCRAB109878.0005
4-Chlorphenylboronsäure ≥98%
4-Chlorphenylboronsäure

Purity: 98+% CAS No.: 1679-18-1 Molecular Formula: C6H6BClO2 Formula Weight: 156.38 Melting Point: 268-277° Beilstein Reference: 2936346

Formel: C₆H₆BClO₂
Molecular Weight: 156.38 g/mol
Schmelzpunkt: 268…277 °C
Dichte: ∼1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039137
CAS-Nummer: 1679-18-1
EINECS: 216-845-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR