4-Chlorphenylboronsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
4-Chlorbenzolboronsäure, Dihydroxy(4-chlorphenyl)boran, (4-Chlorphenyl)metaborsäure
FL10986.POR
010986-100G
010986-25G
010986-5G
CHMPFL10986.POREA
0
EUR
CHMPFL10986.POR
FLUO010986-100G
FLUO010986-25G
FLUO010986-5G
4-Chlorphenylboronsäure
4-Chlorphenylboronsäure
Formel:
C₆H₆BClO₂ Molecular Weight: 156,38 g/mol Schmelzpunkt: 268…272 °C Dichte: ∼1 g/cm³ (20 °C) |
MDL:
MFCD00039137 CAS-Nummer: 1679-18-1 EINECS: 216-845-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...