Pentachlorbenzonitril

Lieferant: CHEMPUR

BTB09647.1 BTB09647.10
CHMPBTB09647.1EA 0 EUR
CHMPBTB09647.1 CHMPBTB09647.10
Pentachlorbenzonitril
Pentachlorbenzonitril
Formel: C₇Cl₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD00173666
CAS-Nummer: 20925-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR