4-Chlor-3-nitrobenzonitril
Lieferant: CHEMPUR
CD07968.1
CD07968.10
CD07968.50
CHMPCD07968.1EA
0
EUR
CHMPCD07968.1
CHMPCD07968.10
CHMPCD07968.50
4-Chlor-3-nitrobenzonitril
4-Chlor-3-nitrobenzonitril
Formel:
C₇H₃ClN₂O₂ Molecular Weight: 182.57 g/mol Schmelzpunkt: 100 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016987 CAS-Nummer: 939-80-0 EINECS: 213-364-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...