4-Chlor-3-nitrobenzonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
380059-25G
380059-5G
SIAL380059-25GEA
88
EUR
SIAL380059-25G
SIAL380059-5G
4-Chlor-3-nitrobenzonitril, Sigma-Aldrich®
4-Chlor-3-nitrobenzonitril
Formel:
C₇H₃ClN₂O₂ Molecular Weight: 182,57 g/mol Schmelzpunkt: 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016987 CAS-Nummer: 939-80-0 EINECS: 213-364-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...