2-Chlor-4-nitrobenzoylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Chlor-4-nitrobenzol-1-carbonylchlorid, 2-Chlor-4-nitrobenzoesäurechlorid

L08246.18 L08246.06
L08246.18EA 186 EUR
L08246.18 L08246.06
2-Chlor-4-nitrobenzoylchlorid ≥98%
2-Chlor-4-nitrobenzoylchlorid

Purity: 98% CAS No.: 7073-36-1 Molecular Formula: C7H3Cl2NO3 Formula Weight: 220.01 Refractive Index: 1.5950 Boiling Point: 165°/20mm UN No.: 3265 MDL No.: MFCD00051515

Formel: C₇H₃Cl₂NO₃
Molecular Weight: 220,01 g/mol
Siedepunkt: 165 °C (20 mmHg)
Schmelzpunkt: 115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051515
CAS-Nummer: 7073-36-1
EINECS: 230-367-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR