4-Chlor-3-nitrobenzoylchlorid

Lieferant: FLUOROCHEM

023908-100G 023908-50G
FLUO023908-100GEA 0 EUR
FLUO023908-100G FLUO023908-50G
4-Chlor-3-nitrobenzoylchlorid
4-Chlor-3-nitrobenzoylchlorid
Formel: C₇H₃Cl₂NO₃
Molecular Weight: 220.01 g/mol
Siedepunkt: 199…200 °C (60 mmHg)
Schmelzpunkt: 51
MDL: MFCD00035743
CAS-Nummer: 38818-50-7
EINECS: 254-133-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR