2-Chlor-4-nitrobenzamid

Lieferant: Thermo Scientific

Synonyme: Aklomid

TL00834DA TL00834EA TL00834.50
MAYBTL00834DAEA 0 EUR
MAYBTL00834DA MAYBTL00834EA MAYBTL00834.50
2-Chlor-4-nitrobenzamid
2-Chlor-4-nitrobenzamid
Formel: C₇H₅ClN₂O₃
MDL: MFCD00017119
CAS-Nummer: 3011-89-0
EINECS: 221-143-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR