3-Chlorbenzhydrazid

Lieferant: Thermo Scientific

Synonyme: 3-Chlorobenzoic hydrazide

SPB06425.1 SPB06425.10 SPB06425.50
MAYBSPB06425.1EA 0 EUR
MAYBSPB06425.1 MAYBSPB06425.10 MAYBSPB06425.50
3-Chlorbenzhydrazid
3-Chlorbenzhydrazid
Formel: C₇H₇ClN₂O
Molecular Weight: 170.6 g/mol
MDL: MFCD00014757
CAS-Nummer: 1673-47-8
EINECS: 216-812-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR