4-Chlorbenzhydrazid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chlorobenzoic hydrazide

A11911.06 A11911.18 A11911.22
A11911.06EA 26.4 EUR
A11911.06 A11911.18 A11911.22
4-Chlorbenzhydrazid ≥98%
4-Chlorbenzhydrazid

Purity: 98% CAS No.: 536-40-3 Molecular Formula: C7H7ClN2O Formula Weight: 170.60 Melting Point: 162-166° Beilstein Reference: 511154

Formel: C₇H₇ClN₂O
Molecular Weight: 170.6 g/mol
Siedepunkt: 354
Schmelzpunkt: 162…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007603
CAS-Nummer: 536-40-3
EINECS: 208-632-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR