4-Chlorbenzhydrazid
Lieferant: Thermo Scientific
Synonyme:
4-Chlorobenzoic hydrazide
SPB05520.1
SPB05520EA
MAYBSPB05520.1EA
0
EUR
MAYBSPB05520.1
MAYBSPB05520EA
4-Chlorbenzhydrazid
4-Chlorbenzhydrazid
Formel:
C₇H₇ClN₂O Molecular Weight: 170.6 g/mol Siedepunkt: 354 Schmelzpunkt: 164 |
MDL:
MFCD00007603 CAS-Nummer: 536-40-3 EINECS: 208-632-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...