3-Chlor-4-methoxybenzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Chlor-p-anissäure

A10106.06 A10106.14
A10106.06EA 54.4 EUR
A10106.06 A10106.14
3-Chlor-4-methoxybenzoesäure ≥98%
3-Chlor-4-methoxybenzoesäure

Purity: 98+% CAS No.: 37908-96-6 Molecular Formula: C8H7ClO3 Formula Weight: 186.59 Melting Point: 215-219° Beilstein Reference: 2087584

Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
Schmelzpunkt: 215…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016512
CAS-Nummer: 37908-96-6
EINECS: 253-708-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR