3-Chlor-5-methoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

696374-1G 696374-5G
SIAL696374-1GEA 46.1 EUR
SIAL696374-1G SIAL696374-5G
3-Chlor-5-methoxybenzoesäure, Sigma-Aldrich®
3-Chlor-5-methoxybenzoesäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD01317994
CAS-Nummer: 82477-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR