3-Chlor-2,6-difluorbenzaldehyd

Lieferant: FLUOROCHEM
Achtung

005587-5G 005587-1G
FLUO005587-5GEA 97.1 EUR
FLUO005587-5G FLUO005587-1G
3-Chlor-2,6-difluorbenzaldehyd
3-Chloro-2,6-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
Molecular Weight: 176.55 g/mol
MDL: MFCD01631323
CAS-Nummer: 190011-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR