3-Chlor-2-fluorbenzaldehyd
Lieferant: FLUOROCHEM
|
Achtung
|
005613-5G
005613-10G
005613-25G
FLUO005613-5GEA
31.1
EUR
FLUO005613-5G
FLUO005613-10G
FLUO005613-25G
3-Chlor-2-fluorbenzaldehyd
3-Chloro-2-fluorobenzaldehyde
Formel:
C₇H₄ClFO |
MDL:
MFCD01631571 CAS-Nummer: 85070-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...