2-Chlor-3,6-difluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-3,6-difluorbenzol-1-carbonylchlorid
005593-5G
005593-10G
FLUO005593-5GEA
0
EUR
FLUO005593-5G
FLUO005593-10G
2-Chlor-3,6-difluorbenzoylchlorid
2-Chlor-3,6-difluorbenzoylchlorid
Formel:
C₇H₂Cl₂F₂O Siedepunkt: 222 °C (1013 hPa) Schmelzpunkt: 70 °C |
MDL:
MFCD01631442 CAS-Nummer: 261762-42-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...