2-Chlor-4-fluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-4-fluorbenzol-1-carbonylchlorid, 2-Chlor-4-fluorbenzoesäurechlorid
004438-5G
004438-25G
FLUO004438-5GEA
42.9
EUR
FLUO004438-5G
FLUO004438-25G
2-Chlor-4-fluorbenzoylchlorid
2-Chlor-4-fluorbenzoylchlorid
Formel:
C₇H₃Cl₂FO Molecular Weight: 193 g/mol |
MDL:
MFCD00084954 CAS-Nummer: 21900-54-9 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...