2-Chlor-4-fluorbenzoylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-4-fluorbenzol-1-carbonylchlorid, 2-Chlor-4-fluorbenzoesäurechlorid

M-Clarity™ Qualitätsstufe = MQ100

703958-25G 703958-5G
SIAL703958-25GEA 194 EUR
SIAL703958-25G SIAL703958-5G
2-Chlor-4-fluorbenzoylchlorid, Sigma-Aldrich®
2-Chlor-4-fluorbenzoylchlorid
Formel: C₇H₃Cl₂FO
Molecular Weight: 193 g/mol
MDL: MFCD00084954
CAS-Nummer: 21900-54-9
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR