2-Chlor-5-fluorbenzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-5-fluorbenzol-1-carbonylchlorid

005620-5G 005620-25G
FLUO005620-5GEA 42.9 EUR
FLUO005620-5G FLUO005620-25G
2-Chlor-5-fluorbenzoylchlorid
2-Chlor-5-fluorbenzoylchlorid
Formel: C₇H₃Cl₂FO
Molecular Weight: 193 g/mol
Siedepunkt: 106
MDL: MFCD01631417
CAS-Nummer: 21900-51-6
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR