2-Chlor-6-fluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-6-fluorbenzol-1-carbonylchlorid
004439-5G
004439-25G
004439-100G
FLUO004439-5GEA
42.9
EUR
FLUO004439-5G
FLUO004439-25G
FLUO004439-100G
2-Chlor-6-fluorbenzoylchlorid
2-Chlor-6-fluorbenzoylchlorid
Formel:
C₇H₃Cl₂FO Molecular Weight: 193 g/mol Siedepunkt: 227 Schmelzpunkt: 57 |
MDL:
MFCD00178746 CAS-Nummer: 79455-63-3 EINECS: 279-162-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...