3-Chlor-2-fluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-2-fluorbenzol-1-carbonylchlorid
CFF944.
005621-1G
005621-5G
005621-10G
CHMPCFF944.EA
0
EUR
CHMPCFF944.
FLUO005621-1G
FLUO005621-5G
FLUO005621-10G
3-Chlor-2-fluorbenzoylchlorid
3-Chlor-2-fluorbenzoylchlorid
Formel:
C₇H₃Cl₂FO Molecular Weight: 193 g/mol Siedepunkt: 202…203 °C (1013 hPa) Dichte: 1.451 g/cm³ (20 °C) |
MDL:
MFCD01631548 CAS-Nummer: 85345-76-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...