3-Chlor-4-fluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-4-fluorbenzol-1-carbonylchlorid
007836-25G
007836-5G
FLUO007836-25GEA
219
EUR
FLUO007836-25G
FLUO007836-5G
3-Chlor-4-fluorbenzoylchlorid
3-Chlor-4-fluorbenzoylchlorid
Formel:
C₇H₃Cl₂FO Molecular Weight: 193 g/mol Siedepunkt: 222 |
MDL:
MFCD00236215 CAS-Nummer: 65055-17-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...