4-Chlor-2,5-difluorbenzoylchlorid

Lieferant: CHEMPUR

FL1628.1 001628-5G 001628-25G
CHMPFL1628.1EA 0 EUR
CHMPFL1628.1 FLUO001628-5G FLUO001628-25G
4-Chlor-2,5-difluorbenzoylchlorid
4-Chlor-2,5-difluorbenzoylchlorid
Formel: C₇H₂Cl₂F₂O
MDL: MFCD00236600
CAS-Nummer: 132794-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR