4-Chlor-2-fluorbenzoylchlorid

Lieferant: FLUOROCHEM

009723-5G 009723-25G
FLUO009723-5GEA 84.9 EUR
FLUO009723-5G FLUO009723-25G
4-Chlor-2-fluorbenzoylchlorid
4-Chlor-2-fluorbenzoylchlorid
Formel: C₇H₃Cl₂FO
Molecular Weight: 193 g/mol
Siedepunkt: 114
CAS-Nummer: 394-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR