4-Chlor-3-fluorbenzoylchlorid

Lieferant: FLUOROCHEM

021890-5G 021890-1G
FLUO021890-5GEA 1400 EUR
FLUO021890-5G FLUO021890-1G
4-Chlor-3-fluorbenzoylchlorid
4-Chlor-3-fluorbenzoylchlorid
Formel: C₇H₃Cl₂FO
Molecular Weight: 193.1 g/mol
MDL: MFCD02091532
CAS-Nummer: 160658-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR