2-Chlor-3,6-difluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-3,6-difluorbenzoesäureamid
005588-1G
005588-5G
FLUO005588-1GEA
60.9
EUR
FLUO005588-1G
FLUO005588-5G
2-Chlor-3,6-difluorbenzamid
2-Chlor-3,6-difluorbenzamid
Formel:
C₇H₄ClF₂NO |
MDL:
MFCD01631444 CAS-Nummer: 261762-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...