2-Chlor-4-fluorbenzamid
Lieferant: FLUOROCHEM
004422-1G
004422-5G
FLUO004422-1GEA
97.1
EUR
FLUO004422-1G
FLUO004422-5G
2-Chlor-4-fluorbenzamid
2-Chlor-4-fluorbenzamid
Formel:
C₇H₅ClFNO Molecular Weight: 173.57 g/mol |
MDL:
MFCD00042531 CAS-Nummer: 88578-90-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...