2-Chlor-5-fluorbenzamid
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
2-Chlor-5-fluorbenzoesäureamid
FL5614.POR
005614-25G
005614-5G
CHMPFL5614.POREA
0
EUR
CHMPFL5614.POR
FLUO005614-25G
FLUO005614-5G
2-Chlor-5-fluorbenzamid
2-Chlor-5-fluorbenzamid
Formel:
C₇H₅ClFNO Molecular Weight: 173.57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631465 CAS-Nummer: 88487-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...