2-Chlor-6-fluorbenzamid

Lieferant: CHEMPUR

Synonyme: 2-Chlor-6-fluorbenzoesäureamid

CD04845.250 CD04845.1
CHMPCD04845.250EA 0 EUR
CHMPCD04845.250 MAYBCD04845.1
2-Chlor-6-fluorbenzamid
2-Chlor-6-fluorbenzamid
Formel: C₇H₅ClFNO
MDL: MFCD00055436
CAS-Nummer: 66073-54-9
EINECS: 266-123-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR