2-Chlor-6-fluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-6-fluorbenzoesäureamid
018028-10G
018028-25G
FLUO018028-10GEA
0
EUR
FLUO018028-10G
FLUO018028-25G
2-Chlor-6-fluorbenzamid
2-Chlor-6-fluorbenzamid
Formel:
C₇H₅ClFNO |
MDL:
MFCD00055436 CAS-Nummer: 66073-54-9 EINECS: 266-123-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...