3-Chlor-2-fluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-2-fluorbenzoesäureamid
CFF922.
005615-5G
005615-10G
CHMPCFF922.EA
0
EUR
CHMPCFF922.
FLUO005615-5G
FLUO005615-10G
3-Chlor-2-fluorbenzamid
3-Chlor-2-fluorbenzamid
Formel:
C₇H₅ClFNO |
MDL:
MFCD00042205 CAS-Nummer: 104326-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...