5-Chlor-2-fluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 5-Chlor-2-fluorbenzoesäureamid, 3-Chlor-6-fluorbenzoesäureamid

CFF926. 005617-5G 005617-10G
CHMPCFF926.EA 0 EUR
CHMPCFF926. FLUO005617-5G FLUO005617-10G
5-Chlor-2-fluorbenzamid
5-Chlor-2-fluorbenzamid
Formel: C₇H₅ClFNO
MDL: MFCD01631435
CAS-Nummer: 261762-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR