3-Chlor-2-fluorbenzoesäure
Lieferant: FLUOROCHEM
CFF589.1
004429-1G
004429.5G
CHMPCFF589.1EA
268
EUR
CHMPCFF589.1
FLUO004429-1G
FLUO004429.5G
3-Chlor-2-fluorbenzoesäure
3-Chloro-2-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174,56 g/mol |
MDL:
MFCD00042506 CAS-Nummer: 161957-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...