2,4-Dichlor-5-fluor-3-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

553867-25G 553867-5G
SIAL553867-25GEA 41.2 EUR
SIAL553867-25G SIAL553867-5G
2,4-Dichlor-5-fluor-3-nitrobenzoesäure, Sigma-Aldrich®
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid
Formel: C₇H₂Cl₂FNO₄
Molecular Weight: 254 g/mol
MDL: MFCD03427137
CAS-Nummer: 106809-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR