1-Chlor-2-fluor-5-iodbenzol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Chlor-4-fluoriodbenzol, 2-Chlor-1-fluor-4-iodbenzol
A11808.14
A11808.22
A11808.14EA
95
EUR
A11808.14
A11808.22
1-Chlor-2-fluor-5-iodbenzol ≥98%
1-Chlor-2-fluor-5-iodbenzol
Purity: 98%
CAS No.: 156150-67-3
Molecular Formula: C6H3ClFI
Formula Weight: 256.45
Flash Point:
Formel:
C₆H₃ClFI Molecular Weight: 256,45 g/mol Siedepunkt: 94…95 °C (15 mmHg) Dichte: 2,020 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042210 CAS-Nummer: 156150-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...